Assay Details
Target Gene Details
Entrez Gene ID: | 100130887 |
Gene Name: | ATE1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121928312-121951965 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATE1-AS1 | NR_120495.1 | |||
| AI634505.1 | ||||
| BF434328.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3814 | Chr.10:121945257 - 121946880 on Build GRCh38 | Loss |
|

Genomic Map