Assay Details
Target Gene Details
Entrez Gene ID: | 92565 |
Gene Name: | fibronectin type III and ankyrin repeat domains 1 |
Gene Aliases: |
HSD13 |
Location: |
Chr.10:125896539-126009599 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANK1 | NM_145235.3 | NP_660278.3 | ||
| XM_005270280.3 | XP_005270337.1 | |||
| XM_011540349.2 | XP_011538651.1 | |||
| XM_011540350.2 | XP_011538652.1 | |||
| XM_011540360.2 | XP_011538662.1 | |||
| XM_011540361.2 | XP_011538663.1 | |||
| XM_011540363.2 | XP_011538665.1 | |||
| XM_017016917.1 | XP_016872406.1 | |||
| XM_017016919.1 | XP_016872408.1 | |||
| XM_017016920.1 | XP_016872409.1 | |||
| XM_017016921.1 | XP_016872410.1 | |||
| XM_017016922.1 | XP_016872411.1 | |||
| XM_017016923.1 | XP_016872412.1 | |||
| XM_017016924.1 | XP_016872413.1 | |||
| XM_017016925.1 | XP_016872414.1 | |||
| AK304407.1 | ||||
| AY358154.1 | AAQ88521.1 | |||
| BC024189.2 | AAH24189.2 | |||
| BQ225468.1 | ||||
| CR627249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762884 | Chr.10:125925704 - 125969535 on Build GRCh38 | Loss |
|
| esv2761638 | Chr.10:125892202 - 126013242 on Build GRCh38 | Loss |
|
| nsv527133 | Chr.10:125945140 - 125967498 on Build GRCh38 | Loss |
|
| esv2759788 | Chr.10:125765331 - 126098133 on Build GRCh38 | Gain |
|

Genomic Map