Assay Details
Target Gene Details
Entrez Gene ID: | 80013 |
Gene Name: | family with sequence similarity 188 member A |
Gene Aliases: |
C10orf97, CARP, DERP5, MST126, MSTP126, my042 |
Location: |
Chr.10:15778169-15860533 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM188A | NM_001318330.1 | NP_001305259.1 | ||
| NM_024948.3 | NP_079224.1 | |||
| XM_005252600.2 | XP_005252657.1 | |||
| XM_005252602.2 | XP_005252659.1 | |||
| XM_006717508.2 | XP_006717571.1 | |||
| XM_011519690.2 | XP_011517992.1 | |||
| XM_011519691.2 | 14 | 2942 | XP_011517993.1 | |
| XM_011519693.1 | XP_011517995.1 | |||
| XM_011519694.1 | XP_011517996.1 | |||
| XM_017016671.1 | XP_016872160.1 | |||
| XM_017016672.1 | XP_016872161.1 | |||
| XM_017016673.1 | XP_016872162.1 | |||
| XM_017016674.1 | XP_016872163.1 | |||
| XM_017016675.1 | XP_016872164.1 | |||
| XM_017016676.1 | XP_016872165.1 | |||
| AB014761.1 | BAB87802.1 | |||
| AF063600.1 | AAG43159.1 | |||
| AF176916.1 | AAQ13660.1 | |||
| AK023459.1 | BAB14582.1 | |||
| AL833873.1 | CAD38730.1 | |||
| BC020605.1 | AAH20605.1 | |||
| BC067799.1 | AAH67799.1 | |||
| CR457315.1 | CAG33596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831795 | Chr.10:15683863 - 15832969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map