Assay Details
Target Gene Details
Entrez Gene ID: | 120526 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C24 |
Gene Aliases: |
DPH4, JJJ3, ZCSL3 |
Location: |
Chr.11:31369829-31432835 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC24 | NM_181706.4 | NP_859057.4 | ||
| XM_017017216.1 | XP_016872705.1 | |||
| AK123087.1 | ||||
| AK289665.1 | ||||
| AL833128.1 | ||||
| CD685768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744312 | Chr.11:31405912 - 31408743 on Build GRCh38 | Deletion |
|
| nsv832106 | Chr.11:31217866 - 31408189 on Build GRCh38 | Gain |
|
| esv3625828 | Chr.11:31375534 - 31451834 on Build GRCh38 | Gain |
|
| nsv1046329 | Chr.11:31185798 - 31502655 on Build GRCh38 | Loss |
|
| esv2751022 | Chr.11:31041408 - 31526385 on Build GRCh38 | Loss |
|
| nsv529004 | Chr.11:30992197 - 31700936 on Build GRCh38 | Loss |
|
| nsv553949 | Chr.11:31296288 - 31518040 on Build GRCh38 | Loss |
|
| nsv1040432 | Chr.11:31293973 - 31411280 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map