Assay Details
Target Gene Details
Entrez Gene ID: | 51400 |
Gene Name: | protein phosphatase methylesterase 1 |
Gene Aliases: |
PME-1 |
Location: |
Chr.11:74170891-74254703 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPME1 | NM_001271593.1 | NP_001258522.1 | ||
| NM_016147.2 | NP_057231.1 | |||
| XM_017017913.1 | XP_016873402.1 | |||
| AF157028.1 | AAD44976.1 | |||
| AK001381.1 | BAA91661.1 | |||
| AK022725.1 | ||||
| AK225647.1 | ||||
| BC003046.1 | AAH03046.1 | |||
| BC050705.2 | AAH50705.1 | |||
| DA691258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34979 | Chr.11:74167643 - 74822407 on Build GRCh38 | Gain |
|
| esv2759837 | Chr.11:74131564 - 74846552 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map