Assay Details
Target Gene Details
Entrez Gene ID: | 338699 |
Gene Name: | ankyrin repeat domain 42 |
Gene Aliases: |
PPP1R79, SARP |
Location: |
Chr.11:83193739-83256099 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD42 | NM_001300972.1 | NP_001287901.1 | ||
| NM_001300973.1 | NP_001287902.1 | |||
| NM_001300975.1 | NP_001287904.1 | |||
| NM_182603.3 | NP_872409.2 | |||
| AK095193.1 | BAC04496.1 | |||
| DQ508815.1 | ||||
| DQ508934.1 | ||||
| EF041819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33531 | Chr.11:82520885 - 83805659 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140319201] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map