Assay Details
Target Gene Details
Entrez Gene ID: | 159989 |
Gene Name: | deuterosome assembly protein 1 |
Gene Aliases: |
CCDC67 |
Location: |
Chr.11:93329079-93438470 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEUP1 | NM_181645.3 | NP_857596.2 | ||
| XM_005273802.1 | XP_005273859.1 | |||
| XM_011542629.2 | XP_011540931.1 | |||
| XM_011542630.1 | XP_011540932.1 | |||
| XM_011542631.2 | XP_011540933.1 | |||
| XM_011542632.2 | XP_011540934.1 | |||
| XM_011542633.2 | XP_011540935.1 | |||
| XM_011542635.2 | XP_011540937.1 | |||
| XM_011542637.2 | XP_011540939.1 | |||
| XM_017017245.1 | XP_016872734.1 | |||
| AK058122.1 | BAB71673.1 | |||
| AK127973.1 | BAC87213.1 | |||
| BC017929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039134 | Chr.11:93056834 - 93486776 on Build GRCh38 | Loss |
|
| nsv468793 | Chr.11:93334997 - 93396727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map