Assay Details
Target Gene Details
Entrez Gene ID: | 196294 |
Gene Name: | inner mitochondrial membrane peptidase subunit 1 |
Gene Aliases: |
IMMP1, IMP1, IMP1-LIKE |
Location: |
Chr.11:31432399-31509644 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMMP1L | NM_001304274.1 | NP_001291203.1 | ||
| NM_144981.2 | NP_659418.1 | |||
| XM_005252812.3 | XP_005252869.1 | |||
| XM_011519942.1 | XP_011518244.1 | |||
| XM_011519943.2 | XP_011518245.1 | |||
| XM_011519944.2 | XP_011518246.1 | |||
| XM_011519945.2 | XP_011518247.1 | |||
| XM_011519946.2 | XP_011518248.1 | |||
| XM_011519947.2 | XP_011518249.1 | |||
| XM_017017305.1 | XP_016872794.1 | |||
| XM_017017306.1 | XP_016872795.1 | |||
| XM_017017307.1 | XP_016872796.1 | |||
| AK057788.1 | BAB71573.1 | |||
| AK310496.1 | ||||
| AK311611.1 | ||||
| BC023595.2 | AAH23595.1 | |||
| DB288287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1740n54 | Chr.11:31433428 - 31674788 on Build GRCh38 | Loss |
|
| nsv1046329 | Chr.11:31185798 - 31502655 on Build GRCh38 | Loss |
|
| esv2751022 | Chr.11:31041408 - 31526385 on Build GRCh38 | Loss |
|
| nsv529004 | Chr.11:30992197 - 31700936 on Build GRCh38 | Loss |
|
| nsv553949 | Chr.11:31296288 - 31518040 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map