Assay Details
Target Gene Details
Entrez Gene ID: | 3854 |
Gene Name: | keratin 6B |
Gene Aliases: |
CK-6B, CK6B, K6B, KRTL1, PC2, PC4 |
Location: |
Chr.12:52446651-52452126 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT6B | NM_005555.3 | NP_005546.2 | ||
| AK296729.1 | ||||
| BC034535.1 | AAH34535.1 | |||
| BG679927.1 | ||||
| L42612.1 | AAC41771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983325 | Chr.12:52446620 - 52452276 on Build GRCh38 | Duplication |
|
| esv2751106 | Chr.12:52434655 - 52470456 on Build GRCh38 | Loss |
|
| nsv1039931 | Chr.12:52443831 - 52473994 on Build GRCh38 | Gain |
|
| dgv1504n100 | Chr.12:52423017 - 52454608 on Build GRCh38 | Gain |
|
| nsv832410 | Chr.12:52321596 - 52506980 on Build GRCh38 | Gain |
|
| nsv1035936 | Chr.12:52444922 - 52504209 on Build GRCh38 | Gain |
|
| nsv832411 | Chr.12:52347803 - 52494080 on Build GRCh38 | Loss |
|
| nsv832412 | Chr.12:52390206 - 52545452 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map