Assay Details
Target Gene Details
Entrez Gene ID: | 3178 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A1 |
Gene Aliases: |
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1 |
Location: |
Chr.12:54280690-54287088 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA1 | NR_135167.1 | |||
| XM_005268826.1 | XP_005268883.1 | |||
| BC070315.1 | AAH70315.1 | |||
| X79536.1 | CAA56072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049160 | Chr.12:54254119 - 54362754 on Build GRCh38 | Gain |
|
| nsv832420 | Chr.12:54137272 - 54296797 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map