Assay Details
Target Gene Details
Entrez Gene ID: | 23141 |
Gene Name: | ankyrin repeat and LEM domain containing 2 |
Gene Aliases: |
KIAA0692, LEMD7, Lem4, MCPH16 |
Location: |
Chr.12:132725667-132761888 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKLE2 | NM_015114.2 | 13 | 4258 | NP_055929.1 |
| XM_005266159.3 | 13 | 4179 | XP_005266216.1 | |
| XM_005266160.1 | 13 | 4115 | XP_005266217.1 | |
| XM_006719735.1 | 12 | 3577 | XP_006719798.1 | |
| XM_017019076.1 | 14 | 4198 | XP_016874565.1 | |
| AB014592.1 | 12 | 3705 | BAA31667.1 | |
| AK024061.1 | 8 | 2946 | ||
| AK025933.1 | 3 | 1953 | ||
| BC062373.1 | 3 | 2266 | AAH62373.1 | |
| BC082962.1 | 3 | 2266 | AAH82962.1 | |
| R59085.1 | 1 | 242 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047500 | Chr.12:132666737 - 132763400 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
| nsv561121 | Chr.12:132717747 - 132729651 on Build GRCh38 | Gain |
|
| esv3631311 | Chr.12:132713141 - 132737944 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map