Assay Details
Target Gene Details
Entrez Gene ID: | 84318 |
Gene Name: | coiled-coil domain containing 77 |
Gene Aliases: |
- |
Location: |
Chr.12:389350-442642 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC77 | NM_001130146.1 | NP_001123618.1 | ||
| NM_001130147.1 | NP_001123619.1 | |||
| NM_001130148.1 | NP_001123620.1 | |||
| NM_032358.3 | NP_115734.1 | |||
| AK027638.1 | BAB55255.1 | |||
| AK293165.1 | ||||
| AK308216.1 | ||||
| AK316029.1 | ||||
| BC006444.2 | AAH06444.1 | |||
| CX784327.1 | ||||
| DA711064.1 | ||||
| DC331633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv514668 | Chr.12:397308 - 412366 on Build GRCh38 | Gain |
|
| esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
|
| nsv1050781 | Chr.12:212187 - 674733 on Build GRCh38 | Loss |
|
| nsv509448 | Chr.12:408807 - 432443 on Build GRCh38 | Insertion |
|
| esv3892121 | Chr.12:386051 - 528197 on Build GRCh38 | Gain |
|
| nsv442250 | Chr.12:323642 - 434681 on Build GRCh38 | Gain |
|
| nsv571 | Chr.12:407796 - 450481 on Build GRCh38 | Insertion |
|

Genomic Map