Assay Details
Target Gene Details
Entrez Gene ID: | 101929384 |
Gene Name: | uncharacterized LOC101929384 |
Gene Aliases: |
- |
Location: |
Chr.12:203645-205093 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929384 | NR_120475.1 |
Target Gene Details
Entrez Gene ID: | 6539 |
Gene Name: | solute carrier family 6 member 12 |
Gene Aliases: |
BGT-1, BGT1, GAT2 |
Location: |
Chr.12:190077-214523 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A12 | NM_001122847.2 | NP_001116319.1 | ||
| NM_001122848.2 | NP_001116320.1 | |||
| NM_001206931.1 | NP_001193860.1 | |||
| NM_003044.4 | NP_003035.3 | |||
| XM_005253747.4 | XP_005253804.1 | |||
| XM_005253748.4 | XP_005253805.1 | |||
| XM_006719005.3 | XP_006719068.1 | |||
| XM_011521010.1 | XP_011519312.1 | |||
| XM_017019841.1 | XP_016875330.1 | |||
| AK125026.1 | ||||
| AK131564.1 | BAD18697.1 | |||
| AK307167.1 | ||||
| AK313690.1 | ||||
| BC126215.1 | ||||
| BC126217.1 | ||||
| L42300.1 | AAA66574.1 | |||
| U27699.1 | AAA87029.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035539 | Chr.12:119253 - 224079 on Build GRCh38 | Gain |
|
| nsv556855 | Chr.12:154673 - 226756 on Build GRCh38 | Loss |
|
| dgv1331n100 | Chr.12:148770 - 207194 on Build GRCh38 | Gain |
|
| nsv556861 | Chr.12:184347 - 251448 on Build GRCh38 | Loss |
|
| nsv832302 | Chr.12:57392 - 234835 on Build GRCh38 | Gain |
|
| nsv556856 | Chr.12:170292 - 218255 on Build GRCh38 | Loss |
|
| esv3628226 | Chr.12:157297 - 251505 on Build GRCh38 | Gain |
|
| esv3628221 | Chr.12:114421 - 232803 on Build GRCh38 | Gain |
|
| nsv523142 | Chr.12:189877 - 223694 on Build GRCh38 | Gain |
|
| dgv1330n100 | Chr.12:45001 - 233978 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map