Assay Details
Target Gene Details
Entrez Gene ID: | 6540 |
Gene Name: | solute carrier family 6 member 13 |
Gene Aliases: |
GAT-2, GAT2, GAT3 |
Location: |
Chr.12:220621-262930 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A13 | NM_001190997.2 | NP_001177926.1 | ||
| NM_016615.4 | NP_057699.2 | |||
| XM_011521012.2 | XP_011519314.1 | |||
| XM_011521013.1 | XP_011519315.1 | |||
| XM_011521014.1 | XP_011519316.1 | |||
| XM_017019843.1 | XP_016875332.1 | |||
| XM_017019844.1 | XP_016875333.1 | |||
| XM_017019846.1 | XP_016875335.1 | |||
| XM_017019847.1 | XP_016875336.1 | |||
| AK295904.1 | ||||
| AK296127.1 | ||||
| AK296209.1 | ||||
| AK313511.1 | ||||
| BC022392.1 | AAH22392.1 | |||
| DA374290.1 | ||||
| U76343.2 | AAF64247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556861 | Chr.12:184347 - 251448 on Build GRCh38 | Loss |
|
| esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
|
| nsv1050781 | Chr.12:212187 - 674733 on Build GRCh38 | Loss |
|
| nsv556857 | Chr.12:170292 - 268738 on Build GRCh38 | Gain |
|
| esv3628226 | Chr.12:157297 - 251505 on Build GRCh38 | Gain |
|
| esv3628228 | Chr.12:240958 - 244359 on Build GRCh38 | Loss |
|
| nsv470253 | Chr.12:218255 - 250297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map