Assay Details
Target Gene Details
Entrez Gene ID: | 55652 |
Gene Name: | solute carrier family 48 member 1 |
Gene Aliases: |
HRG-1, HRG1, hHRG-1 |
Location: |
Chr.12:47753605-47782753 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC48A1 | XM_005269016.4 | XP_005269073.1 | ||
| XM_011538558.2 | XP_011536860.1 | |||
| XM_017019612.1 | XP_016875101.1 | |||
| XM_017019613.1 | XP_016875102.1 | |||
| XM_017019614.1 | XP_016875103.1 | |||
| XM_017019615.1 | XP_016875104.1 | |||
| XM_017019616.1 | XP_016875105.1 | |||
| BC002759.2 | AAH02759.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470289 | Chr.12:47716768 - 47786725 on Build GRCh38 | Gain |
|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map