Assay Details
Target Gene Details
Entrez Gene ID: | 55297 |
Gene Name: | coiled-coil domain containing 91 |
Gene Aliases: |
HSD8, p56 |
Location: |
Chr.12:28190427-28550166 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC91 | XM_005253413.1 | XP_005253470.1 | ||
| XM_006719104.2 | XP_006719167.1 | |||
| XM_006719105.1 | XP_006719168.1 | |||
| XM_011520729.2 | XP_011519031.1 | |||
| XM_011520734.1 | XP_011519036.1 | |||
| XM_011520735.1 | XP_011519037.1 | |||
| XM_017019569.1 | XP_016875058.1 | |||
| XM_017019570.1 | XP_016875059.1 | |||
| XM_017019571.1 | XP_016875060.1 | |||
| XM_017019572.1 | XP_016875061.1 | |||
| XM_017019574.1 | XP_016875063.1 | |||
| XM_017019575.1 | XP_016875064.1 | |||
| XM_017019576.1 | XP_016875065.1 | |||
| XM_017019577.1 | XP_016875066.1 | |||
| AK093152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557864 | Chr.12:27899261 - 28277169 on Build GRCh38 | Gain |
|
| nsv557876 | Chr.12:27961883 - 28572171 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map