Assay Details
Target Gene Details
Entrez Gene ID: | 23329 |
Gene Name: | TBC1 domain family member 30 |
Gene Aliases: |
- |
Location: |
Chr.12:64759489-64881022 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D30 | XM_017019085.1 | XP_016874574.1 | ||
| XM_017019086.1 | XP_016874575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2682n54 | Chr.12:64609189 - 64806142 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map