Assay Details
Target Gene Details
Entrez Gene ID: | 23141 |
Gene Name: | ankyrin repeat and LEM domain containing 2 |
Gene Aliases: |
KIAA0692, LEMD7, Lem4, MCPH16 |
Location: |
Chr.12:132725667-132761888 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKLE2 | NM_015114.2 | 4 | 1022 | NP_055929.1 |
| XM_005266159.3 | 4 | 943 | XP_005266216.1 | |
| XM_005266160.1 | 4 | 879 | XP_005266217.1 | |
| XM_006719735.1 | 4 | 933 | XP_006719798.1 | |
| XM_011534787.2 | 4 | 933 | XP_011533089.1 | |
| XM_011534788.1 | 4 | 933 | XP_011533090.1 | |
| XM_017019076.1 | 4 | 879 | XP_016874565.1 | |
| AB014592.1 | 3 | 469 | BAA31667.1 | |
| BC001530.2 | 3 | 311 | AAH01530.2 | |
| BC025347.1 | 3 | 311 | AAH25347.1 | |
| BC043157.1 | 4 | 968 | AAH43157.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631312 | Chr.12:132735138 - 132794488 on Build GRCh38 | Loss |
|
| nsv561129 | Chr.12:132736267 - 132769626 on Build GRCh38 | Loss |
|
| esv3631313 | Chr.12:132742571 - 132754625 on Build GRCh38 | Loss |
|
| nsv1047500 | Chr.12:132666737 - 132763400 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map