Assay Details
Target Gene Details
Entrez Gene ID: | 400165 |
Gene Name: | ATP11A upstream neighbor |
Gene Aliases: |
C13orf35, SMABLO1 |
Location: |
Chr.13:112647044-112684497 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP11AUN | NM_207440.2 | NP_997323.1 | ||
| AK129953.1 | BAC85256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv563226 | Chr.13:112629877 - 112688892 on Build GRCh38 | Gain |
|
| dgv3417n54 | Chr.13:112504873 - 112744470 on Build GRCh38 | Gain |
|
| nsv1045238 | Chr.13:112530283 - 112723969 on Build GRCh38 | Gain |
|
| nsv1044644 | Chr.13:112437023 - 113055517 on Build GRCh38 | Gain |
|

Genomic Map