Assay Details
Target Gene Details
Entrez Gene ID: | 100505538 |
Gene Name: | RBM26 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:79406309-79424336 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM26-AS1 | NR_038991.1 | |||
| AK002196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523979 | Chr.13:79405544 - 79546128 on Build GRCh38 | Loss |
|
| esv3632819 | Chr.13:79393219 - 79532503 on Build GRCh38 | Gain |
|
| nsv832657 | Chr.13:79330843 - 79524973 on Build GRCh38 | Gain |
|
| esv3632814 | Chr.13:79199135 - 79520919 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs137982247] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map