Assay Details
Target Gene Details
Entrez Gene ID: | 79774 |
Gene Name: | growth hormone regulated TBC protein 1 |
Gene Aliases: |
TBC1D6 |
Location: |
Chr.13:113324164-113364158 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRTP1 | NM_001286732.1 | NP_001273661.1 | ||
| NM_001286733.1 | NP_001273662.1 | |||
| NM_024719.3 | NP_078995.2 | |||
| XM_005268333.3 | XP_005268390.1 | |||
| XM_011537521.2 | XP_011535823.1 | |||
| XM_017020755.1 | XP_016876244.1 | |||
| XM_017020756.1 | XP_016876245.1 | |||
| AB449887.1 | ||||
| AK026127.1 | BAB15368.1 | |||
| AK300897.1 | ||||
| BC033071.2 | AAH33071.2 | |||
| BC062461.1 | AAH62461.1 | |||
| BC080640.1 | AAH80640.1 | |||
| BC112123.1 | AAI12124.1 | |||
| BC112125.1 | AAI12126.1 | |||
| BQ689859.1 | ||||
| CX788865.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951927 | Chr.13:113312286 - 113380185 on Build GRCh38 | Deletion |
|
| nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain |
|
| nsv509521 | Chr.13:113308960 - 113394480 on Build GRCh38 | Insertion |
|
| nsv563316 | Chr.13:113261171 - 113347907 on Build GRCh38 | Loss |
|
| nsv563318 | Chr.13:113319297 - 113430196 on Build GRCh38 | Loss |
|
| nsv563317 | Chr.13:113292168 - 113369086 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map