Assay Details
Target Gene Details
Entrez Gene ID: | 220081 |
Gene Name: | glutamate rich 6B |
Gene Aliases: |
FAM194B |
Location: |
Chr.13:45541203-45615739 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH6B | NM_182542.2 | NP_872348.2 | ||
| XM_011534965.2 | XP_011533267.1 | |||
| XM_017020418.1 | XP_016875907.1 | |||
| AK057244.1 | BAB71393.1 | |||
| BC131494.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| nsv510583 | Chr.13:45484456 - 45586401 on Build GRCh38 | Deletion |
|

Genomic Map