Assay Details
Target Gene Details
Entrez Gene ID: | 10901 |
Gene Name: | dehydrogenase/reductase 4 |
Gene Aliases: |
CR, NRDR, PHCR, PSCD, SCAD-SRL, SDR-SRL, SDR25C1, SDR25C2 |
Location: |
Chr.14:23953735-23969279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS4 | NM_001282987.1 | NP_001269916.1 | ||
| NM_001282988.1 | NP_001269917.1 | |||
| NM_001282989.1 | NP_001269918.1 | |||
| NM_001282990.1 | NP_001269919.1 | |||
| NM_001282991.1 | NP_001269920.1 | |||
| NM_021004.3 | NP_066284.2 | |||
| XM_006720005.3 | XP_006720068.1 | |||
| XM_011536366.2 | XP_011534668.1 | |||
| AB045131.1 | BAB18775.1 | |||
| AF044127.1 | AAD02292.1 | |||
| AF064256.1 | AAQ13444.1 | |||
| AK001870.1 | BAA91953.1 | |||
| AK301201.1 | ||||
| AK308436.1 | ||||
| AK314448.1 | ||||
| AY071856.1 | AAL61824.2 | |||
| AY358638.1 | AAQ89001.1 | |||
| AY616182.1 | AAT70757.1 | |||
| BC003019.1 | AAH03019.1 | |||
| BP366505.1 | ||||
| BU529016.1 | ||||
| CD674505.1 | ||||
| DQ325464.1 | ABC61320.1 | |||
| DQ344810.1 | ABD75823.1 |
Target Gene Details
Entrez Gene ID: | 55449 |
Gene Name: | DHRS4 antisense RNA 1 |
Gene Aliases: |
AS1DHRS4, C14orf167, DHRS4AS1, PRO1488 |
Location: |
Chr.14:23938731-23955112 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS4-AS1 | NR_023921.2 | 1 | 176 | |
| NR_023922.2 | ||||
| NR_023923.2 | ||||
| NR_023924.2 | 1 | 176 | ||
| BF206895.1 | ||||
| BX447332.2 | 1 | 101 | ||
| BX482101.1 | ||||
| BX647964.1 | ||||
| DA439818.1 | 1 | 176 | ||
| DB254318.1 | ||||
| HQ845197.1 | ||||
| HQ845198.1 | ||||
| HQ845199.1 | ||||
| JQ979294.1 | ||||
| KJ545484.1 | ||||
| KJ574270.1 | ||||
| KJ574271.1 | ||||
| KJ574272.1 | ||||
| KJ574273.1 | 1 | 93 | ||
| KJ574274.1 | 1 | 139 | ||
| KJ599456.1 | 1 | 126 | ||
| KJ599459.1 | ||||
| KJ599460.1 | 1 | 170 | ||
| KJ599461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1217 | Chr.14:23931356 - 24043480 on Build GRCh38 | Deletion |
|
| nsv564046 | Chr.14:23943563 - 24011124 on Build GRCh38 | Loss |
|
| nsv1218 | Chr.14:23941636 - 23983409 on Build GRCh38 | Insertion |
|
| esv3633790 | Chr.14:23953691 - 23970340 on Build GRCh38 | Loss |
|
| dgv1838n100 | Chr.14:23945076 - 24031780 on Build GRCh38 | Gain |
|
| nsv89 | Chr.14:23935989 - 24006004 on Build GRCh38 | Deletion |
|
| esv27040 | Chr.14:23951116 - 23954459 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map