Assay Details
Target Gene Details
Entrez Gene ID: | 123722 |
Gene Name: | fibronectin type III and SPRY domain containing 2 |
Gene Aliases: |
SPRYD1 |
Location: |
Chr.15:82755365-82807698 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSD2 | NM_001007122.3 | 13 | 5883 | NP_001007123.1 |
| NM_001281805.1 | 13 | 5865 | NP_001268734.1 | |
| NM_001281806.1 | 12 | 5748 | NP_001268735.1 | |
| AL833295.1 | 13 | 5883 |
Target Gene Details
Entrez Gene ID: | 677778 |
Gene Name: | small Cajal body-specific RNA 15 |
Gene Aliases: |
ACA45 |
Location: |
Chr.15:82755945-82756071 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARNA15 | NR_003011.1 | |||
| AJ609485.1 |
Target Gene Details
Entrez Gene ID: | 100505616 |
Gene Name: | small nucleolar RNA host gene 21 |
Gene Aliases: |
FSD2-AS1, SCARNA15HG |
Location: |
Chr.15:82750564-82757206 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNHG21 | NR_110096.1 | |||
| NR_110097.1 | ||||
| NR_110098.1 | ||||
| AA284521.1 | ||||
| BX112407.1 | ||||
| CF129490.1 | ||||
| CK904397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469557 | Chr.15:82749011 - 82891877 on Build GRCh38 | Loss |
|
| dgv2637n100 | Chr.15:82568764 - 82803755 on Build GRCh38 | Gain |
|
| nsv471687 | Chr.15:82749012 - 82891877 on Build GRCh38 | Gain |
|
| nsv817702 | Chr.15:82630710 - 82788672 on Build GRCh38 | Loss |
|
| esv34318 | Chr.15:82614804 - 82784743 on Build GRCh38 | Loss |
|
| esv2674021 | Chr.15:82628241 - 82789040 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73443066] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map