Assay Details
Target Gene Details
Entrez Gene ID: | 11173 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 7 |
Gene Aliases: |
ADAM-TS 7, ADAM-TS7, ADAMTS-7 |
Location: |
Chr.15:78759203-78811464 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS7 | NM_014272.4 | NP_055087.2 | ||
| XM_005254137.3 | XP_005254194.1 | |||
| XM_011521166.2 | XP_011519468.1 | |||
| AK299844.1 | ||||
| AL110226.1 | CAB53684.1 | |||
| AY327122.1 | ||||
| BC061631.1 | AAH61631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952004 | Chr.15:78730159 - 78800258 on Build GRCh38 | Deletion |
|
| esv33573 | Chr.15:78761142 - 79001539 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148106161] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map