Assay Details
Target Gene Details
Entrez Gene ID: | 80031 |
Gene Name: | semaphorin 6D |
Gene Aliases: |
- |
Location: |
Chr.15:47184101-47774223 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6D | NM_001198999.1 | NP_001185928.1 | ||
| XM_011522075.2 | XP_011520377.1 | |||
| XM_017022620.1 | XP_016878109.1 | |||
| XM_017022621.1 | XP_016878110.1 | |||
| AB040912.2 | ||||
| BC150253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670559 | Chr.15:47191101 - 47195595 on Build GRCh38 | Deletion |
|
| esv33210 | Chr.15:47185332 - 47312112 on Build GRCh38 | Gain |
|
| esv3636383 | Chr.15:47191130 - 47195580 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map