Assay Details
Target Gene Details
Entrez Gene ID: | 197003 |
Gene Name: | uncharacterized protein MGC15885 |
Gene Aliases: |
- |
Location: |
Chr.15:62637172-62645181 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGC15885 | NR_026897.1 |
Target Gene Details
Entrez Gene ID: | 83660 |
Gene Name: | talin 2 |
Gene Aliases: |
ILWEQ |
Location: |
Chr.15:62390527-62844631 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLN2 | XM_005254708.4 | XP_005254765.1 | ||
| XM_005254710.4 | XP_005254767.1 | |||
| XM_005254711.4 | XP_005254768.1 | |||
| XM_005254712.4 | XP_005254769.1 | |||
| XM_005254713.4 | XP_005254770.1 | |||
| XM_005254714.3 | XP_005254771.1 | |||
| XM_005254715.2 | XP_005254772.1 | |||
| XM_006720717.3 | XP_006720780.1 | |||
| XM_017022665.1 | XP_016878154.1 | |||
| XM_017022666.1 | XP_016878155.1 | |||
| XM_017022667.1 | XP_016878156.1 | |||
| XM_017022668.1 | XP_016878157.1 | |||
| XM_017022669.1 | XP_016878158.1 | |||
| AB621808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044132 | Chr.15:62434402 - 62650249 on Build GRCh38 | Gain |
|
| nsv1035441 | Chr.15:62406037 - 63349997 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map