Assay Details
Target Gene Details
Entrez Gene ID: | 146212 |
Gene Name: | potassium channel tetramerization domain containing 19 |
Gene Aliases: |
- |
Location: |
Chr.16:67289490-67326758 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD19 | NM_001100915.1 | NP_001094385.1 | ||
| AK097481.1 | BAC05072.1 | |||
| AK302567.1 | ||||
| AK302748.1 | ||||
| BC117335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv572906 | Chr.16:67237955 - 67368685 on Build GRCh38 | Loss |
|
| nsv471092 | Chr.16:67174030 - 67518392 on Build GRCh38 | Gain |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs538392855] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map