Assay Details
Target Gene Details
Entrez Gene ID: | 1453 |
Gene Name: | casein kinase 1 delta |
Gene Aliases: |
ASPS, CKIdelta, FASPS2, HCKID |
Location: |
Chr.17:82239317-82273742 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1D | NM_001893.4 | NP_001884.2 | ||
| NM_139062.2 | NP_620693.1 | |||
| NR_110578.1 | ||||
| XM_005256336.3 | XP_005256393.1 | |||
| XM_005256337.4 | XP_005256394.1 | |||
| XM_017024199.1 | XP_016879688.1 | |||
| AB091044.1 | BAC10903.1 | |||
| AB209463.1 | BAD92700.1 | |||
| AK291758.1 | ||||
| AK295638.1 | ||||
| AK296969.1 | ||||
| AK303365.1 | ||||
| AK308852.1 | ||||
| AK311248.1 | ||||
| BC003558.1 | AAH03558.1 | |||
| BC015775.1 | AAH15775.1 | |||
| U29171.1 | AAC50807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
|
| nsv833572 | Chr.17:82204977 - 82388835 on Build GRCh38 | Loss |
|
| esv2762577 | Chr.17:82244836 - 82838431 on Build GRCh38 | Gain+Loss |
|
| esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73366234,rs73366236] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map