Assay Details
Target Gene Details
Entrez Gene ID: | 440419 |
Gene Name: | TBC1 domain family member 3 pseudogene 5 |
Gene Aliases: |
- |
Location: |
Chr.17:27418005-27431623 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D3P5 | NR_033892.1 | |||
| BC112354.1 | AAI12355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv442414 | Chr.17:27417246 - 27430467 on Build GRCh38 | Loss |
|
| esv22166 | Chr.17:27416057 - 27429589 on Build GRCh38 | Gain+Loss |
|
| nsv960080 | Chr.17:27411872 - 27433976 on Build GRCh38 | Duplication |
|
| dgv540e199 | Chr.17:26964108 - 27464626 on Build GRCh38 | Deletion |
|
| esv2760459 | Chr.17:27417133 - 27430463 on Build GRCh38 | Loss |
|
| nsv2014 | Chr.17:27391938 - 27437190 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map