Assay Details
Target Gene Details
Entrez Gene ID: | 55288 |
Gene Name: | ras homolog family member T1 |
Gene Aliases: |
ARHT1, MIRO-1, MIRO1 |
Location: |
Chr.17:32142454-32253374 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640377 | Chr.17:32099879 - 32211610 on Build GRCh38 | Gain |
|
| esv2758684 | Chr.17:31894702 - 32210963 on Build GRCh38 | Gain |
|
| nsv1066680 | Chr.17:32059518 - 32197864 on Build GRCh38 | Gain |
|
| nsv1057055 | Chr.17:32104914 - 32210956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map