Assay Details
Target Gene Details
Entrez Gene ID: | 100128006 |
Gene Name: | uncharacterized LOC100128006 |
Gene Aliases: |
- |
Location: |
Chr.17:12760134-12790284 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128006 | NR_104607.1 | |||
| BX649083.1 | ||||
| DA231808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34095 | Chr.17:12436261 - 12841981 on Build GRCh38 | Loss |
|
| esv3582530 | Chr.17:12775230 - 12785568 on Build GRCh38 | Loss |
|

Genomic Map