Assay Details
Target Gene Details
Entrez Gene ID: | 286753 |
Gene Name: | tumor suppressor candidate 5 |
Gene Aliases: |
DSPB1, IFITMD3, LOST1 |
Location: |
Chr.17:1279663-1300987 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUSC5 | NM_172367.2 | NP_758955.2 | ||
| AB090231.1 | BAC43751.1 | |||
| AL831929.1 | ||||
| BC142615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761939 | Chr.17:1262192 - 1322646 on Build GRCh38 | Loss |
|
| dgv3081n100 | Chr.17:1095450 - 1360904 on Build GRCh38 | Gain |
|
| nsv509645 | Chr.17:1249608 - 1334590 on Build GRCh38 | Insertion |
|
| dgv3080n100 | Chr.17:987971 - 1330752 on Build GRCh38 | Gain |
|
| nsv952097 | Chr.17:1257607 - 1308606 on Build GRCh38 | Deletion |
|
| nsv1064006 | Chr.17:1189272 - 1339670 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map