Assay Details
Target Gene Details
Entrez Gene ID: | 6147 |
Gene Name: | ribosomal protein L23a |
Gene Aliases: |
L23A, MDA20 |
Location: |
Chr.17:28719982-28724356 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL23A | NM_000984.5 | NP_000975.2 | ||
| AK130965.1 | ||||
| AK312123.1 | ||||
| BC058041.1 | AAH58041.1 | |||
| BG476195.1 | ||||
| BP382007.1 | ||||
| L13799.1 | AAA35681.1 | |||
| U02032.1 | AAA03341.1 | |||
| U43701.1 | AAB03210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
| nsv960457 | Chr.17:28720734 - 28723723 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map