Assay Details
Target Gene Details
Entrez Gene ID: | 339201 |
Gene Name: | ASB16 antisense RNA 1 |
Gene Aliases: |
C17orf65 |
Location: |
Chr.17:44175973-44186717 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB16-AS1 | NR_049729.1 | |||
| NR_049730.1 | ||||
| AK055685.1 | ||||
| CB151608.1 | ||||
| CN367999.1 | ||||
| DA567080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833455 | Chr.17:44108553 - 44297300 on Build GRCh38 | Loss |
|
| dgv529n67 | Chr.17:44170904 - 44246444 on Build GRCh38 | Gain |
|

Genomic Map