Assay Details
Target Gene Details
Entrez Gene ID: | 146880 |
Gene Name: | Rho GTPase activating protein 27 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.17:64749662-64781999 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC146880 | NR_026899.1 | |||
| AK055342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066765 | Chr.17:64520760 - 65128185 on Build GRCh38 | Gain |
|
| dgv3274n100 | Chr.17:64618099 - 64978067 on Build GRCh38 | Gain |
|
| nsv469875 | Chr.17:64741770 - 64944984 on Build GRCh38 | Loss |
|
| dgv44n68 | Chr.17:64726315 - 64900956 on Build GRCh38 | Loss |
|
| nsv960140 | Chr.17:64749454 - 64758635 on Build GRCh38 | Duplication |
|

Genomic Map