Assay Details
Target Gene Details
Entrez Gene ID: | 10230 |
Gene Name: | neighbor of BRCA1 gene 2 (non-protein coding) |
Gene Aliases: |
NCRNA00192 |
Location: |
Chr.17:43125583-43145113 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBR2 | NR_003108.1 | |||
| AL598622.1 | ||||
| BC107773.1 | ||||
| CR936675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv575053 | Chr.17:43039112 - 43146838 on Build GRCh38 | Loss |
|
| esv2672913 | Chr.17:42964983 - 43322347 on Build GRCh38 | Deletion |
|
| nsv470587 | Chr.17:43069579 - 43146837 on Build GRCh38 | Loss |
|

Genomic Map