Assay Details
Target Gene Details
Entrez Gene ID: | 10939 |
Gene Name: | AFG3 like matrix AAA peptidase subunit 2 |
Gene Aliases: |
SCA28, SPAX5 |
Location: |
Chr.18:12328944-12377309 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFG3L2 | NM_006796.2 | NP_006787.2 | ||
| XM_011525601.2 | XP_011523903.1 | |||
| AK129504.1 | ||||
| BC024282.1 | AAH24282.1 | |||
| BC065016.1 | AAH65016.1 | |||
| Y18314.1 | CAB48398.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060808 | Chr.18:12302793 - 12514418 on Build GRCh38 | Gain |
|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map