Assay Details
Target Gene Details
Entrez Gene ID: | 23253 |
Gene Name: | ankyrin repeat domain 12 |
Gene Aliases: |
ANCO-2, ANCO1, GAC-1, Nbla00144 |
Location: |
Chr.18:9136753-9285985 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 23253 |
Gene Name: | ankyrin repeat domain 12 |
Gene Aliases: |
ANCO-2, ANCO1, GAC-1, Nbla00144 |
Location: |
Chr.18:9136753-9285985 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |