Assay Details
Target Gene Details
Entrez Gene ID: | 7561 |
Gene Name: | zinc finger protein 14 |
Gene Aliases: |
GIOT-4, KOX6 |
Location: |
Chr.19:19710472-19733112 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF14 | NM_021030.2 | NP_066358.2 | ||
| AB021644.1 | BAA86990.1 | |||
| AK074832.1 | ||||
| BC136323.1 | ||||
| BM836276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3455n100 | Chr.19:19695577 - 19758523 on Build GRCh38 | Gain |
|
| esv33937 | Chr.19:19716057 - 19720846 on Build GRCh38 | Loss |
|
| nsv2436 | Chr.19:19705049 - 19758362 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map