Assay Details
Target Gene Details
Entrez Gene ID: | 440503 |
Gene Name: | perilipin 5 |
Gene Aliases: |
LSDA5, LSDP5, MLDP, OXPAT |
Location: |
Chr.19:4522531-4535196 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLIN5 | NM_001013706.2 | NP_001013728.2 | ||
| BC131524.1 | ||||
| DQ839131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833720 | Chr.19:4367721 - 4540556 on Build GRCh38 | Loss |
|
| nsv578444 | Chr.19:4502189 - 4559634 on Build GRCh38 | Gain |
|
| nsv428361 | Chr.19:4510042 - 4689189 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map