Assay Details
Target Gene Details
Entrez Gene ID: | 404665 |
Gene Name: | CACTIN antisense RNA 1 |
Gene Aliases: |
C19orf29-AS1, C19orf29OS |
Location: |
Chr.19:3607247-3613930 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACTIN-AS1 | NR_038865.1 | |||
| AK097999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv833713 | Chr.19:3564423 - 3627361 on Build GRCh38 | Loss |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73529990] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map