Assay Details
Target Gene Details
Entrez Gene ID: | 148268 |
Gene Name: | zinc finger protein 570 |
Gene Aliases: |
- |
Location: |
Chr.19:37467465-37488652 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF570 | NM_001300993.2 | 6 | 4850 | NP_001287922.1 |
| NM_001321991.1 | 5 | 4699 | NP_001308920.1 | |
| NM_001321992.1 | 4 | 5030 | NP_001308921.1 | |
| NM_001321993.1 | 5 | 5173 | NP_001308922.1 | |
| NM_001321994.1 | 4 | 5089 | NP_001308923.1 | |
| NM_144694.3 | 5 | 5114 | NP_653295.1 | |
| BM671447.1 | 1 | 485 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056753 | Chr.19:37472812 - 37525041 on Build GRCh38 | Loss |
|
| nsv833819 | Chr.19:37408599 - 37560697 on Build GRCh38 | Gain |
|
| nsv579468 | Chr.19:37486707 - 37802847 on Build GRCh38 | Gain |
|
| esv2758759 | Chr.19:37303948 - 37560704 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map