Assay Details
Target Gene Details
Entrez Gene ID: | 388514 |
Gene Name: | cytochrome P450 family 4 subfamily F member 24, pseudogene |
Gene Aliases: |
- |
Location: |
Chr.19:15759334-15779988 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4F24P | NR_033864.1 | 10 | 1868 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060711 | Chr.19:15745103 - 15817596 on Build GRCh38 | Gain |
|
| esv3334671 | Chr.19:15673766 - 15931354 on Build GRCh38 | Duplication |
|
| nsv833766 | Chr.19:15759095 - 15956904 on Build GRCh38 | Loss |
|
| nsv1064684 | Chr.19:15311319 - 16062011 on Build GRCh38 | Gain |
|
| esv2718244 | Chr.19:15743561 - 15951200 on Build GRCh38 | Deletion |
|

Genomic Map