Assay Details
Target Gene Details
Entrez Gene ID: | 6874 |
Gene Name: | TATA-box binding protein associated factor 4 |
Gene Aliases: |
TAF2C, TAF2C1, TAF4A, TAFII130, TAFII135 |
Location: |
Chr.20:61974798-62065810 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF4 | NM_003185.3 | NP_003176.2 | ||
| AK097744.1 | 1 | 865 | ||
| AK123937.1 | ||||
| U75308.1 | AAC50901.1 | |||
| Y11354.1 | CAA72189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568094 | Chr.20:61973373 - 61984015 on Build GRCh38 | Loss |
|
| nsv955146 | Chr.20:61694945 - 62020244 on Build GRCh38 | Deletion |
|
| nsv509781 | Chr.20:61911923 - 62050571 on Build GRCh38 | Insertion |
|
| nsv428378 | Chr.20:61886206 - 62008187 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73297753] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map