Assay Details
Target Gene Details
Entrez Gene ID: | 105372836 |
Gene Name: | uncharacterized LOC105372836 |
Gene Aliases: |
- |
Location: |
Chr.21:45055869-45073903 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105372836 | XM_011529812.2 | XP_011528114.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|

Genomic Map