Assay Details
Target Gene Details
Entrez Gene ID: | 728299 |
Gene Name: | keratin associated protein 19-8 |
Gene Aliases: |
- |
Location: |
Chr.21:31038159-31038476 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP19-8 | NM_001099219.1 | 1 | 203 | NP_001092689.1 |
| AB096964.1 | 1 | 203 | BAE46379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587361 | Chr.21:30990059 - 31071890 on Build GRCh38 | Gain |
|
| esv3575385 | Chr.21:31000408 - 31807461 on Build GRCh38 | Gain |
|
| esv2762107 | Chr.21:30956893 - 31183479 on Build GRCh38 | Gain |
|

Genomic Map