Assay Details
Target Gene Details
Entrez Gene ID: | 164714 |
Gene Name: | tubulin tyrosine ligase like 8 |
Gene Aliases: |
- |
Location: |
Chr.22:50018575-50058227 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL8 | XM_017029176.1 | XP_016884665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829329 | Chr.22:49941791 - 50034475 on Build GRCh38 | Gain |
|
| dgv849e214 | Chr.22:50013481 - 50024531 on Build GRCh38 | Gain |
|
| nsv1151614 | Chr.22:50019170 - 50023871 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map