Assay Details
Target Gene Details
Entrez Gene ID: | 100873985 |
Gene Name: | MED14 opposite strand |
Gene Aliases: |
MED14-AS1 |
Location: |
Chr.X:40735396-40738701 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED14OS | NM_001289773.1 | 2 | 771 | NP_001276702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525223 | Chr.X:40664921 - 40754042 on Build GRCh38 | Gain |
|
| nsv518965 | Chr.X:40569250 - 40754042 on Build GRCh38 | Gain |
|

Genomic Map