Assay Details
Target Gene Details
Entrez Gene ID: | 8573 |
Gene Name: | calcium/calmodulin dependent serine protein kinase |
Gene Aliases: |
CAGH39, CAMGUK, CMG, FGS4, LIN2, MICPCH, MRXSNA, TNRC8 |
Location: |
Chr.X:41514934-41923525 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASK | NM_001126054.2 | NP_001119526.1 | ||
| NM_001126055.2 | NP_001119527.1 | |||
| NM_003688.3 | NP_003679.2 | |||
| XM_005272686.4 | XP_005272743.1 | |||
| XM_006724566.3 | XP_006724629.1 | |||
| XM_011543993.2 | XP_011542295.1 | |||
| XM_011543994.2 | XP_011542296.1 | |||
| XM_011543995.2 | XP_011542297.1 | |||
| XM_011543996.2 | XP_011542298.1 | |||
| AB208859.1 | BAD92096.1 | |||
| AF032119.1 | AAB88125.1 | |||
| AF035582.1 | AAB88198.1 | |||
| AF262405.1 | AAF72667.1 | |||
| AK313399.1 | ||||
| AY705392.1 | AAU10527.1 | |||
| BC117311.1 | ||||
| BC143454.1 | ||||
| BC143456.1 | ||||
| DN993605.1 |
Target Gene Details
Entrez Gene ID: | 27197 |
Gene Name: | G protein-coupled receptor 82 |
Gene Aliases: |
- |
Location: |
Chr.X:41724155-41730135 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR82 | NM_080817.4 | 3 | 2233 | NP_543007.1 |
| XM_017029414.1 | 2 | 2389 | XP_016884903.1 | |
| AK122626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33720 | Chr.X:40891487 - 43157518 on Build GRCh38 | Loss |
|
| nsv523134 | Chr.X:41600467 - 41917489 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map